
Citrinin 98%
Catalog # ACRO229580050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder or flakes
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:250.25 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,II
:GHS06
CAS Number:518-75-2
Formula:C₁₃H₁₄O₅
MDL Number:MFCD00006912