Protein L agarose, Pierce™
Pierce™ Protein L agarose is a premium quality Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
- Immobilised Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein L is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
- Standard capacity beads with a normal load of immobilised Protein L
Binding capacity of 5 to 10 mg human IgG/ml resin