Immunoassay microplates, anti-GST coated, 96-well strip plates, Pierce™
Catalog # PIER15144
Supplier: Thermo Fisher Scientific
Specifications
- Packed:5 plates per pack
- Colour:Clear
- Description:96-well strip plate, PS, anti-GST coated
- Pk:5
Specifications
About this item
Pierce™ Anti-GST coated plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use.
- Antibody-based - plates are coated with mouse monoclonal anti-GST antibody
- Sensitive - detects GST fusion proteins down to 7,5 ng/well
- Flexibility - binds native or denatured forms of GST; an alternative to glutathione-coated plates
- Ready to use - pre-blocked with SuperBlock Blocking Buffer
- Compatibility - Thermo Scientific Cell Lysis Reagents and many other popular lysis buffers do not interfere with GST-fusion protein binding
- Superior performance - improved signal-to-noise ratio relative to competitor's plates
Unlike glutathione coated plates, these anti-GST plates effectively bind both native and denatured forms of GST. In most cases, pre-purification of cell lysates is not necessary before screening and analysis of recombinant GST-tagged protein expression by ELISA using the plates.
Binding property: standard capacity, affinity
Detection method: colorimetric
Surface treatment: antibody