HyClone™ Calf Serum, U.S. Origin
Supplier: HyClone products (Cytiva)
Certificates
About this item
HyClone™ bovine calf serum can be used as an effective alternative to foetal bovine serum (FBS).
- Filtered through sequential 100 nm pore size rated filters
- Extensive quality, performance and analytical testing
- Endotoxin Concentration ≤ 10 EU/mL
- Storage Temperature is –10 °C
Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.
Certifications: Fully traceable and conforms to all USDA regulations.
Specifications
- Catalog No:
- HYCLSH30073.03
- HYCLSH30073.02
- HYCLSH30073.04
- SH30073.03IR25
- HYCLSH30073.02HI
- HYCLSH30073.04HI
- SH30073.02IR
- HYCLSH30073.03HI
- Origin:
- US
- Endotoxin:
- ≤10 EU/ml
- Haemoglobin:
- <30 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.