Immobilised Streptavidin resin, UltraLink™, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Pierce™ Streptavidin UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilised onto a beaded acrylamide support. UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
- Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Streptavidin UltraLink™ resin binding capacity: >2 mg of biotinylated BSA/ml of resin
Streptavidin Plus UltraLink™ resin binding capacity: >4 mg of biotinylated BSA/ml of resin