Order Entry
Finland
ContactUsLinkComponent
E.Z.N.A.® Plant DNA DS kit
E.Z.N.A.® Plant DNA DS kit
Supplier:  OMEGA BIO-TEK
Certificates
E.Z.N.A.® Plant DNA DS kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Sample size:
    50 mg of wet tissue or 10 mg dry tissue
  • Description:
    E.Z.N.A.® Plant DNA DS Kit

Specifications

Related Information