Order Entry
Finland
ContactUsLinkComponent
Subcellular Protein Fractionation Kit for Cultured Cells
Subcellular Protein Fractionation Kit for Cultured Cells
Catalog # 78840.
CAS Number:  
undefined
Subcellular Protein Fractionation Kit for Cultured Cells
Catalog # 78840.
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Subcellular protein fractionation kit for cultured cells, sufficient for 50 samples each containing 2 million cells (20 µl packed)
Specifications
About this item
This kit is ideal for segregation and enrichment of proteins from five different cellular compartments.
  • Concise - extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours
  • Convenient - perform a simple procedure without using ultracentrifugation over gradients
  • Complete - obtain protein fractions from a single kit
  • Cultured cells - developed and optimized for use with cultured mammalian cells
  • Compatible - use extracts for downstream applications such as protein assays, Western blotting, protein assays, electrophoretic mobility shift assays, reporter-gene and enzyme-activity assays
Delivery: Kit includes four extraction buffers, a stabilised nuclease and halt protease inhibitor cocktail.