Epoprosterol sodium ≥99%, white powder
Catalog # ICNA0215196901
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
Specification Test Results
About this item
:
:374.453 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:61849-14-7
Formula:C₂₀H₃₁NaO₅
MDL Number:MFCD00135629