0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
D-Gulonic acid-γ-lactone
D-Gulonic acid-γ-lactone, powder
D-Gulonic acid-γ-lactone, powder
D-Gulonic acid-γ-lactone, powder
Supplier:
MP Biomedicals
Certificates
Expand image
D-Gulonic acid-γ-lactone, powder
Supplier:
MP Biomedicals
About this item
CAS Number:
6322-07-2
:
Formula:
C₆H₁₀O₆
MDL Number:
MFCD00005391
:
178.14 g/mol