Samplers, PowderThief
Supplier: Sampling Systems
Certificates
About this item
These PowderThief samplers are suitable for sampling free flowing powders and granules.
- Made of high quaility 316 stainless steel
- PTFE used for clutch mechanism
- Single point sampler
A range of tips are available so that a single sampler can be used to accurately sample a range of different volumes.