NMR tubes, high quality grade, KIMBLE®
Supplier: DWK Life Sciences
Certificates
About this item
Borosilicate glass, 51 expansion, Type I and ASTM E438, Type I, Class B.
- With attached polyethylene caps
- These HQ NMR tubes are highest quality grade and are 100% gauged for wall thickness, concentricity and camber specifications.
External diameter: 10 mm
Internal diameter: 9,07 mm
Wall thickness: 0,46 mm