Order Entry
Finland
ContactUsLinkComponent
Test kit, Phosphorus TNTplus®
Test kit, Phosphorus TNTplus®
Catalog # HACHTNT845
Supplier:  Hach
CAS Number:  
undefined
Test kit, Phosphorus TNTplus®
Catalog # HACHTNT845
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Ascorbic acid
  • Pk:
    1
  • Description:
    Phosphorus TNT plus kit, reactive and total, UHR
  • Seq:
    03
  • Range:
    2 - 20 mg/l
Specifications
About this item
TNT Plus kit is used for the efficient and accurate determination of reactive (ortho) and total phosphorus (phosphate) using TNTplus® vials.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary