Order Entry
Finland
Orders LinkContactUsLinkComponent
Polysorbate 40 NF

Polysorbate 40 NF

Supplier:   Spectrum Chemical
Polysorbate 40 NF
Supplier:  Spectrum Chemical
Certificates
Polysorbate 40 NF

3 Options Available Below

Product Details & Documents
Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass, amber
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 667,00 €
    667,00 €
    1 * 4 L
       667,00 €

  • Poly drum
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L

  • 200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L - 10 380,00 €
    10 380,00 €
    1 * 200 L
       10 380,00 €