Order Entry
Finland
Orders LinkContactUsLinkComponent
Ethanolamine NF

Ethanolamine NF

Corrosive
Irritant
Supplier:   Spectrum Chemical
Ethanolamine NF
Supplier:  Spectrum Chemical
Certificates
Ethanolamine NF

3 Options Available Below

Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:13,03762
  • Boiling Pt:171 °C (1013 hPa)
  • Melting Pt:10.5 °C
  • Density:1.0164 g/cm³ (20 °C)
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Assay:
    98% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass, amber
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 1 470,00 €
    1 470,00 €
    1 * 4 L
       1 470,00 €

  • Poly drum
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L

  • 200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L