Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethylene glycol-lithium acetate
Polyethylene glycol-lithium acetate solution
Polyethylene glycol-lithium acetate solution
Polyethylene glycol-lithium acetate solution
Catalog # J62213.AK
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
Polyethylene glycol-lithium acetate solution
Catalog # J62213.AK
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
250 mL
Seq:
2590
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Refrigerator
Product Details
Documentation