Order Entry
Finland
ContactUsLinkComponent
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
Irritant
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
Catalog # L17433.06
CAS Number:  
Irritant
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 98%
Catalog # L17433.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:145100-50-1
 :
Formula:C₇H₄F₆N₂O₄S₂
MDL Number:MFCD00191834
 :358.23 g/mol
Boiling Pt:97 °C (2 mmHg)
Melting Pt:37…41 °C
Flash Pt:>110 °C (230 °F)
 :GHS07
Storage Temperature:Refrigerator