Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amberlite® IR-120 (Na)
Amberlite® IR-120 (Na) ion exchange resin
Amberlite® IR-120 (Na) ion exchange resin
Amberlite® IR-120 (Na) ion exchange resin
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Amberlite® IR-120 (Na) ion exchange resin
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
9002-23-7
:
68441-33-8
MDL Number:
MFCD00132707
Merck Index:
14,382
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL15199
Videos
Thermo Scientific Acroseal 25 Years
Documentation