

Propargyl acetate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-09-8
:
Formula:C₅H₆O₂
MDL Number:MFCD00041601
:98.10 g/mol
Boiling Pt:124 °C
Density:0.989
Flash Pt:31 °C (88 °F)
Storage Temperature:Ambient
ADR:3,III
:GHS06
:GHS02