Cyclopropyl phenyl ketone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3481-02-5
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00001294
:146.19 g/mol
Boiling Pt:248 °C
Density:1.06
Flash Pt:90 °C (194 °F)
Storage Temperature:Ambient
EINECS:222-458-2