Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bisindolylmaleimide I
Bisindolylmaleimide I
Bisindolylmaleimide I
Bisindolylmaleimide I
Catalog # J63401.MB
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
Bisindolylmaleimide I
Catalog # J63401.MB
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
Documentation
:
:
412.48 g/mol
CAS Number:
133052-90-1
Formula:
C₂₅H₂₄N₄O₂
MDL Number:
MFCD00236428
Product Details
Documentation