Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dextran sulphate sodium salt
Dextran sulphate sodium salt, M.W. ∼ 8,000
Dextran sulphate sodium salt, M.W. ∼ 8,000
Dextran sulphate sodium salt, M.W. ∼ 8,000
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Dextran sulphate sodium salt, M.W. ∼ 8,000
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
9011-18-1
:
Formula:
(C₆H₇Na₃O₁₄S₃)n
MDL Number:
MFCD00081551
Merck Index:
14,2951
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ62101
Videos
Thermo Scientific Acroseal 25 Years
Documentation