Papain (from Carica Papaya Latex)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Applications: A cysteine protease; breaks down intercellular matrix of cartilage
Specifications
- CAS No.:9001-73-4
- Species:Carica papaya latex
- Storage conditions:2...8 °C
- Form:White to off-white lyophilized powder
- MDL No.:MFCD00131791
- Merck reference:14,7016
- Enzyme name:Papain
- Molecular weight:23 kDa