Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Proline ethyl ester hydrochloride
D-Proline ethyl ester hydrochloride 98%
D-Proline ethyl ester hydrochloride 98%
D-Proline ethyl ester hydrochloride 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
D-Proline ethyl ester hydrochloride 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
131477-20-8
:
Formula:
C₇H₁₄ClNO₂
MDL Number:
MFCD00191003
:
179.644 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation