2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester 96%
Catalog # H62587.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.07 g/mol
Storage Temperature:Ambient
CAS Number:1429340-48-6
Formula:C₁₃H₁₆BFO₄
MDL Number:MFCD23098820