
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:330641-16-2
:
Formula:C₁₁H₁₅BClF₄N₅O
MDL Number:MFCD04973270
:355.53 g/mol
Melting Pt:198…204 °C
:GHS07
Storage Temperature:Ambient