
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 96%
Catalog # H54570.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.10 g/mol
Melting Pt:131…133 °C
:GHS07
Storage Temperature:Ambient
CAS Number:893440-50-1
Formula:C₁₂H₁₉BN₂O₃
MDL Number:MFCD12923427