Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-2-aminothiazole-5-carboxylate
Ethyl-2-aminothiazole-5-carboxylate 97%
Ethyl-2-aminothiazole-5-carboxylate 97%
:
Thermo Fisher Scientific
Ethyl-2-aminothiazole-5-carboxylate 97%
:
Thermo Fisher Scientific
:
32955-21-8
:
:
C₆H₈N₂O₂S
:
MFCD00602139
:
172.21 g/mol
:
158…160 °C
msds_ALFAH31769
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years