Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic
:
Thermo Fisher Scientific
Biphenyl diphenyl ether eutectic
:
Thermo Fisher Scientific
:
8004-13-5
:
:
C₂₄H₂₀O
:
MFCD00148859
:
324.423 g/mol
:
257 °C
:
12…14 °C
:
1.056
:
113 °C (235 °F)
:
Ambient
:
616-827-0
:
3082
:
9,III
:
GHS07
msds_ALFAH30347
Thermo Scientific Acroseal 25 Years