Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Dehydrated Media for Microbiology
YP Base medium
YP Base medium
YP Base medium
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
YP Base medium
Supplier:
Thermo Fisher Scientific
Certificates
About this item
MDL No.: MFCD01636187
Specifications
MDL No.:
MFCD01636187
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH26407
Videos
Thermo Scientific Acroseal 25 Years
Documentation