
8-Mercaptomenthone cis- and trans-mixture 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38462-22-5
:
Formula:C₁₀H₁₈OS
MDL Number:MFCD00012393
:186.31 g/mol
Boiling Pt:270…272 °C
Density:1.00
Flash Pt:108 °C (226 °F)
EINECS:253-953-1
:GHS07
Storage Temperature:Ambient