Nerolidol (cis and trans mixture) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7212-44-4
:
Formula:C₁₅H₂₆O
MDL Number:MFCD00008911
:222.37 g/mol
Merck Index:14,6477
Boiling Pt:263…266 °C
Density:0.876
Flash Pt:96 °C (204 °F)
Storage Temperature:Ambient
EINECS:230-597-5