Ethyl-4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2386-26-7
:
Formula:C₁₁H₁₅NO₃
MDL Number:MFCD00022373
:209.25 g/mol
Melting Pt:140…143 °C
Storage Temperature:Ambient