
1,3-dimethyl-1H-pyrazol-5-amine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3524-32-1
:
Formula:C₅H₉N₃
MDL Number:MFCD00051651
:111.148 g/mol
Melting Pt:72…77 °C
Flash Pt:88 °C (190 °F)
:GHS07
Storage Temperature:Ambient