Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetrakisepoxy cyclosiloxane
Tetrakisepoxy cyclosiloxane
Tetrakisepoxy cyclosiloxane
Irritant
Tetrakisepoxy cyclosiloxane
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Tetrakisepoxy cyclosiloxane
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
257284-60-9
:
Formula:
C₂₈H₅₆O₁₂Si₄
MDL Number:
MFCD22380706
:
697.09 g/mol
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA046960
Videos
Thermo Scientific Acroseal 25 Years
Documentation