Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper etchant
Copper etchant
Copper etchant
Copper etchant
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Copper etchant
Supplier:
Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:
MFCD00161912
Density:
1.19
Storage Temperature:
Ambient
UN:
3264
ADR:
8,III
:
GHS05
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA044583
Videos
Thermo Scientific Acroseal 25 Years
Documentation