Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
C-276 alloy
C-276 alloy, foil, Thickness: 0.2 mm (0.008 in)
C-276 alloy, foil, Thickness: 0.2 mm (0.008 in)
C-276 alloy, foil, Thickness: 0.2 mm (0.008 in)
43994.GH
:
Thermo Fisher Scientific
:
C-276 alloy, foil, Thickness: 0.2 mm (0.008 in)
43994.GH
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
1736
Pk Info:
100×100 mm
:
:
GHS08
:
Ambient
:
MFCD00801065