Copper(II) pyrophosphate hydrate (43.5 - 48.5% P₂O₇)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16570-28-8
Formula:Cu₂P₂O₇·xH₂O
MDL Number:MFCD00150225
:301.02 g/mol (anhy)
EINECS:239-250-2
:GHS07
Storage Temperature:Ambient