Order Entry
Finland
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.25 mm (0.01 in) Type 304
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.25 mm (0.01 in) Type 304
Catalog # 41584.GH
CAS Number:  
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.25 mm (0.01 in) Type 304
Catalog # 41584.GH
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1736
  • Pk Info:
    100×100 mm

Specifications

About this item

 :
 :55.845 g/mol
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499