

Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.025 mm (0.001 in) Type 304
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Fe
MDL Number:MFCD00148499
:55.845 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient