Polyethylene glycol monomethyl ether, M.W. 550
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Melting Pt:18…20 °C
Density:1.10
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient