Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver, brazing alloy
Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 1.6 mm (0.06 in)
Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 1.6 mm (0.06 in)
Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 1.6 mm (0.06 in)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Silver, brazing alloy (Ag:Sn; 3.5:96.5 wt%), Ø 1.6 mm (0.06 in)
Supplier:
Thermo Fisher Scientific
Certificates
About this item
:
Formula:
Ag:Cu:Zn:Sn;56:22:17:5%
MDL Number:
MFCD00801120
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA041032
Videos
Thermo Scientific Acroseal 25 Years
Documentation