

Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.127 mm (0.005 in) Type 304
Catalog # 40944.BZ
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:150 cm
- Seq:7880
Specifications
About this item
:
:55.845 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499