
Zinc 2-ethylhexanoate (∼20% Zn) ≥97% cont. 1% diethylene glycolmonomethyl ether
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:136-53-8
:
Formula:C₁₆H₃₀O₄Zn
MDL Number:MFCD00053316
:351.792 g/mol
EINECS:205-251-1
:GHS08
Storage Temperature:Ambient