Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gallium tin alloy
Gallium tin alloy ≥99.99% (metals basis)
Gallium tin alloy ≥99.99% (metals basis)
:
Thermo Fisher Scientific
Gallium tin alloy ≥99.99% (metals basis)
:
Thermo Fisher Scientific
:
:
Ga:Sn;92:8%
:
MFCD00144390
:
∼25 °C
:
Ambient
:
2803
:
8,III
:
GHS05
msds_ALFA018161
Thermo Scientific Acroseal 25 Years