Bismuth indium lead tin eutectic ingot (alloy 136) ≥99.9% (metals basis)
Catalog # 18143.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:58 °C
:GHS08
Storage Temperature:Ambient
Formula:Bi:In:Pb:Sn;49:21:18:12%
MDL Number:MFCD00210606