Order Entry
Finland
ContactUsLinkComponent
Magnesium aluminum zinc plate (alloy AZ31B), Thickness: 6.35 mm (0.25 in)
Magnesium aluminum zinc plate (alloy AZ31B), Thickness: 6.35 mm (0.25 in)
Catalog # 14066.RF
CAS Number:  
Magnesium aluminum zinc plate (alloy AZ31B), Thickness: 6.35 mm (0.25 in)
Catalog # 14066.RF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Storage Temperature:Ambient
Formula:Mg:Al:Zn;96:3:1 wt%
MDL Number:MFCD01074901