VWR® Kingston, Safety Goggles
Catalog # 111-1841
Supplier: VWR Collection
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Colour:Translucent blue
- Lenses:Clear PC
- Pk:1
Specifications
About this item
Wrap-around modern style goggles, especially designed for small faces, provides excellent panoramic vision. The goggles consist of a PC lens, PVC frame and polyester/rubber headband.
- High comfort with maximum protection
- Fitted with indirect ventilation
- Opalescent frame prevents light reflection
Certifications: Compliant with: EN 166:2001. Frame: EN166 3 4 BT CE. Lens: VWR® 1 BT.