Anti-PRC1 Rabbit Polyclonal Antibody
BSBTPB9814
- Antibody type:Primary
- Antigen name:protein regulator of cytokinesis 1
- Antigen symbol:PRC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9055
- Antigen synonyms:ASE1
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human