Disposable samplers, DispoLance
Supplier: BURKLE
Certificates
About this item
The DispoLance samplers can be used for taking large and small sample quantities quickly and easily, directly from bags.
- Cleanroom manufactured
- Optionally sterilised by gamma rays
- Cross contamination is completely eliminated
Suitable for free-flowing powders and small-grain granulates.