Order Entry
Finland
ContactUsLinkComponent
Surface active cleaning agent, Neutracon®
Surface active cleaning agent, Neutracon®
Catalog # 148-0145
Supplier:  DECON LAB
undefined
Surface active cleaning agent, Neutracon®
Catalog # 148-0145
Supplier:  DECON LAB

Specifications

  • Capacity:
    5 l
  • Description:
    Surface active cleaning agent, Neutracon®
  • Seq:
    02
  • Pk:
    5 L

Specifications

About this item

A surface active cleaning agent and/or decontaminant, for use on materials which may be corroded, etched, discoloured or weakened by acidic or alkaline cleaning agents.

  • Phosphate-free and enzyme-free
  • Non toxic
  • Colourless to pale amber, non viscous liquid which has a faint odour
Certifications: OECD 301E (ISO method 7827-1986 (E))
Packaging: Supplied in 1 and 5 litre HDPE containers, with 'tamper-evident' screw-on caps.
Caution: Not suitable for domestic use nor suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate.